Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFDN6 Antibody (OTI4E10), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273339C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273339C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273339C Supplier Novus Biologicals Supplier No. NBP273339C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PFDN6 Monoclonal specifically detects PFDN6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen PFDN6
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4E10
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias H2-KE2, HLA class II region expressed gene KE2, KE-2, KE2 protein, MGC70744, PFD6HKE2, prefoldin 6, prefoldin subunit 6, Protein Ke2
Gene Symbols PFDN6
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-129 of human PFDN6 (NP_055075) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10471.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.