Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PfHsp40 Rabbit anti-P. falciparum, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB171864 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171864 25 μg
NB171863 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171864 Supplier Novus Biologicals Supplier No. NBP31824025UG
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PfHsp40 Polyclonal antibody specifically detects PfHsp40 in P. falciparum samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen PfHsp40
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1.8 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS pH 7.4, 50% glycerol
Gene Alias DnaJ protein, putative
Host Species Rabbit
Immunogen C-terminal peptide of Pf11_0513 conjugated to KLH
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 811045
Target Species P. falciparum
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.