Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKFB2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232194 Supplier Novus Biologicals Supplier No. NBP232194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PFKFB2 Polyclonal specifically detects PFKFB2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PFKFB2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006203.2
Gene Alias 6PF-2-K/Fru-2,6-P2ase 2, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2, 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2, DKFZp781D2217, fructose-2,6-bisphosphatase, cardiac isozyme, MGC138308,6PF-2-K/Fru-2,6-P2ase heart-type isozyme, MGC138310, PFK/FBPase 2, PFK-2/FBPase-2, PFKFB, cardiac
Gene Symbols PFKFB2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 455 to 505 of human 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 using the numbering given in entry NP_006203.2 (GeneID 5208).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5208
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla and Chimpanzee.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.