Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKFB4 Antibody (OTI1C8), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273340IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273340IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273340IR Supplier Novus Biologicals Supplier No. NBP273340IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PFKFB4 Monoclonal specifically detects PFKFB4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PFKFB4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1C8
Conjugate DyLight 755
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4,6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4,6PF-2-K/Fru-2,6-P2ase 4,6PF-2-K/Fru-2,6-P2ase testis-type isozyme, bifunctional enzyme with kinase and biphosphatase activities, PFK/FBPase 4
Gene Symbols PFKFB4
Host Species Mouse
Immunogen Full length recombinant protein of human PFKFB4 (NP_004558) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5210.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.