Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKP Antibody (OTI1D6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB273344APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273344APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273344APC Supplier Novus Biologicals Supplier No. NBP273344APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PFKP Monoclonal specifically detects PFKP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen PFKP
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone OTI1D6
Conjugate APC
Dilution Flow Cytometry, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias 6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase
Gene Symbols PFKP
Host Species Mouse
Immunogen Full length recombinant protein of human PFKP (NP_002618) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5214.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.