Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFTK1 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB146757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB146757 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB146757 Supplier Novus Biologicals Supplier No. NBP306062IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PFTK1 Polyclonal specifically detects PFTK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PFTK1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cell division protein kinase 14, cyclin-dependent kinase 14, EC 2.7.11, KIAA0834EC 2.7.11.22, PFTAIRE1hPFTAIRE1, PFTK1PFTAIRE protein kinase 1, Serine/threonine-protein kinase PFTAIRE-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PFTK1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5218
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.