Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFTK1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094096 20 μL
NB094095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094096 Supplier Novus Biologicals Supplier No. NBP30367120UL

Rabbit Polyclonal Antibody

PFTK1 Polyclonal specifically detects PFTK1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PFTK1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Cell division protein kinase 14, cyclin-dependent kinase 14, EC 2.7.11, KIAA0834EC 2.7.11.22, PFTAIRE1hPFTAIRE1, PFTK1PFTAIRE protein kinase 1, Serine/threonine-protein kinase PFTAIRE-1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 312-451 of human PFTK1 (NP_036527.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5218
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.