Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP149532Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149532Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149532Z Supplier Novus Biologicals Supplier No. NBP149532AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGAM1 Polyclonal specifically detects PGAM1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen PGAM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B
Gene Symbols PGAM1
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PGAM1 protein (between residues 200-254) [UniProt P18669]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5223
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.