Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM2 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB046669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046669 Supplier Novus Biologicals Supplier No. NBP297695R

Rabbit Polyclonal Antibody

PGAM2 Polyclonal specifically detects PGAM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PGAM2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PGAM2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Proteases & Other Enzymes, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5224
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.