Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGC1 alpha Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N104676J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104676J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N104676J646 Supplier Novus Biologicals Supplier No. NBP104676JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGC1 alpha Polyclonal specifically detects PGC1 alpha in Human, Mouse, Rat, Porcine, Goat, Hamster, Sheep, Squirrel samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.

Specifications

Antigen PGC1 alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha
Gene Symbols PPARGC1A
Host Species Rabbit
Immunogen This PGC1 alpha Antibody was developed against a recombinant protein made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot# Q9UBK2].
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10891.0
Target Species Human, Mouse, Rat, Pig, Goat, Hamster, Sheep, Squirrel
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.