Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGC1 alpha/beta Antibody (SR1068), Novus Biologicals™
SDP

Catalog No. NB260164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260164 50 μg
NB259823 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260164 Supplier Novus Biologicals Supplier No. NBP32170750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PGC1 alpha/beta Monoclonal antibody specifically detects PGC1 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PGC1 alpha/beta
Applications Western Blot
Classification Monoclonal
Clone SR1068
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias 4631412G21Rik;A830037N07Rik;ENSMUSG00000079510;ERRL1;Gm11133;LEM6;PERC;PGC1;PGC-1;PGC-1(alpha);PGC-1(beta);PGC1A;PGC-1alpha;PGC1B;PGC1beta;PGC-1v;Pgco1;PPARGC1;PPARGC1A;PPARGC1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PGC1 alpha/beta (Uniprot #: Q9UBK2, Q86YN6)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.