Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD2 Synthase/PTGDS Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB11059911W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059911W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059911W Supplier Novus Biologicals Supplier No. NB11059911AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGD2 Synthase/PTGDS Polyclonal specifically detects PGD2 Synthase/PTGDS in Human, Mouse, Rat, Bovine, Canine, Feline, Primate, Rabbit samples. It is validated for Western Blot.

Specifications

Antigen PGD2 Synthase/PTGDS
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. B2R727
Gene Alias Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase
Gene Symbols PTGDS
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to region within residue 120-190 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot# P41222]
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5730
Target Species Human, Mouse, Rat, Bovine, Canine, Feline, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.