Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD2 Synthase/PTGDS Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB091091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091091 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091091 Supplier Novus Biologicals Supplier No. NBP300115AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGD2 Synthase/PTGDS Polyclonal specifically detects PGD2 Synthase/PTGDS in Human samples. It is validated for ELISA.

Specifications

Antigen PGD2 Synthase/PTGDS
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PGD2 Synthase/PTGDS (P41222; Met1-Gln190).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5730
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.