Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD2 Synthase/PTGDS Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB331923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331923 Supplier Novus Biologicals Supplier No. NBP297406APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGD2 Synthase/PTGDS Polyclonal antibody specifically detects PGD2 Synthase/PTGDS in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PGD2 Synthase/PTGDS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PGD2 Synthase/PTGDS fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5730
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.