Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGK1 Antibody (2F4), PerCP, Novus Biologicals™
SDP

Catalog No. NB309080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309080 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309080 Supplier Novus Biologicals Supplier No. NBP259392PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGK1 Monoclonal antibody specifically detects PGK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PGK1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2F4
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Cell migration-inducing gene 10 protein, EC 2.7.2.3, MGC142128, MGC8947, MIG10, PGKAMGC117307, phosphoglycerate kinase 1, Primer recognition protein 2, PRP 2
Host Species Mouse
Immunogen Recombinant human PGK1 (1-417aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5230
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.