Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGK2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB325079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325079 Supplier Novus Biologicals Supplier No. NBP299497JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGK2 Polyclonal antibody specifically detects PGK2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PGK2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias dJ417L20.2, EC 2.7.2.3, PGK-2, PGKB, PGKPS, phosphoglycerate kinase 2, phosphoglycerate kinase autosomal pseudogene, Phosphoglycerate kinase, testis specific
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PGK2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5232
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.