Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGL2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB344006 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344006 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344006 Supplier Novus Biologicals Supplier No. NBP241136MFV500

Chicken Polyclonal Antibody

PGL2 Polyclonal antibody specifically detects PGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen PGL2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SDHAF2. The immunogen is located within the first 50 amino acids of SDHAF2. Amino Acid Squence: SFRRFYRGDSPTDSQKD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54949
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.