Learn More
Promega pGL2-Enhancer Vector
Catalog No. PRE1621 Shop All Promega Corporation Products
Click to view available options
Quantity:
20 μg
Includes
  • pGL2-Enhancer DNA
  • Bacterial Strain JM109
Four reporter vectors that contain the coding region for firefly luciferase.
  • Four Reporter Vectors that Contain luc, the Firefly Luciferase Gene
  • All vectors contain the firefly luciferase reporter gene, which enables sensitive and rapid quantitation of reporter activity
  • Deletions and site-directed mutations can be made directly to inserted DNAs without subcloning
  • Upstream polyadenylation signal minimizes spurious transcription of the reporter gene
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.