Learn More
Promega pGL4.83[hRlucP/Puro] Vector
Catalog No. PRE7511 Shop All Promega Corporation Products
Click to view available options
Quantity:
20 μg
Vectors can accept putative promoter elements for investigation or control reporter vectors for dual-luciferase assays. Stably transfected cells can be selected using puromycin.
  • Cloning Vectors For Use as Control Reporter Vectors Offering Different Reporter Protein Stabilities
  • Designed for cloning a putative promoter element for investigating gene transcription control regions
  • Available with three varieties of engineered Renilla luciferase genes: hRluc, hRlucP or hRlucCP
  • Select for stably transfected cells using puromycin
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.