Learn More
PGLYRP1/PGRP-S Antibody (188C424) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | PGLYRP1/PGRP-S |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF |
| Classification | Monoclonal |
| Clone | 188C424 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 2 ug/ml, Flow Cytometry : 1:20-1:20000, Immunohistochemistry : 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000. Use reported in scientific literature (Dukhanina et al (2009)), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Onkelen et al (2012)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 19666596), CyTOF-ready |
| Formulation | PBS |
| Gene Alias | peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L |
| Gene Symbols | PGLYRP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.