Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP1/PGRP-S Antibody (188C424), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N1056719PC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1056719PC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1056719PC7 Supplier Novus Biologicals Supplier No. NB10056719PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGLYRP1/PGRP-S Monoclonal specifically detects PGLYRP1/PGRP-S in Human, Mouse samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen PGLYRP1/PGRP-S
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 188C424
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Gene Symbols PGLYRP1
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8993.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.