Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP2/PGRP-L Antibody (439728), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB303464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303464 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303464 Supplier Novus Biologicals Supplier No. FAB4704T

Rat Monoclonal Antibody

PGLYRP2/PGRP-L Monoclonal antibody specifically detects PGLYRP2/PGRP-L in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen PGLYRP2/PGRP-L
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 439728
Conjugate Alexa Fluor 594
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPLHMFT0141, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L, Ser23-Asn530, Accession # Q8VCS0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 114770
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.