Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGLYRP4/PGRP-I beta Antibody (186C426), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10056721
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).Specifications
PGLYRP4/PGRP-I beta | |
Monoclonal | |
0.5 mg/mL | |
Western Blot, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490) | |
peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta | |
Mouse | |
Protein G purified | |
RUO | |
57115 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
186C426 | |
Unconjugated | |
Q96LB8 | |
PGLYRP4 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta. | |
0.1 mg | |
Primary | |
PGRP-1b (186C426) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction