Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP4/PGRP-I beta Antibody (186C426), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$206.50 - $499.50

Specifications

Antigen PGLYRP4/PGRP-I beta
Clone 186C426
Concentration 0.5 mg/mL
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056721T
SDP
View Documents
Novus Biologicals
NB10056721SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056721
SDP
View Documents
Novus Biologicals
NB10056721
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).
Specifications

Specifications

PGLYRP4/PGRP-I beta
0.5 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry
Unconjugated
Mouse
Human
peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta
PGLYRP4
IgG1
Protein G purified
PGRP-1b (186C426)
186C426
Western Blot, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490)
Monoclonal
Purified
RUO
Q96LB8
57115
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.