Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGLYRP4/PGRP-I beta Antibody (186C426), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106721A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106721A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106721A30 Supplier Novus Biologicals Supplier No. NB10056721AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen PGLYRP4/PGRP-I beta
Applications Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 186C426
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta
Gene Symbols PGLYRP4
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57115
Test Specificity PGRP-1b (186C426)
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.