Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM1 Antibody (84G2), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB161668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161668 Supplier Novus Biologicals Supplier No. NBP259454AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen PGM1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 84G2
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1
Host Species Mouse
Immunogen Recombinant human PGM1 (1-562aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5236
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.