Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGM1 Antibody (CL3299), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PGM1 |
---|---|
Clone | CL3299 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PGM1 Monoclonal specifically detects PGM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PGM1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
5236 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SILATRKQSVEDILKDHWQKHGRNFFTRYDYEEVEAEGANKMMKDLEALMFDRSFVGKQFSANDKVYTVEKADNFEYSD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL3299 | |
Monoclonal | |
Purified | |
Lipid and Metabolism | |
EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1 | |
PGM1 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title