Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM2L1 Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB144455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB144455 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB144455 Supplier Novus Biologicals Supplier No. NBP305946AF405

Rabbit Polyclonal Antibody

PGM2L1 Polyclonal specifically detects PGM2L1 in Human samples. It is validated for Western Blot.

Specifications

Antigen PGM2L1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BM32Aglucose-1,6-bisphosphate synthase, EC 2.7.1, EC 2.7.1.106, FLJ32029, glucose 1,6-bisphosphate synthase, phosphoglucomutase 2-like 1, Phosphoglucomutase-2-like 1, PMMLP
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PGM2L1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 283209
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.