Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM3 Antibody (OTI1A11), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273362C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273362C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273362C Supplier Novus Biologicals Supplier No. NBP273362C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGM3 Monoclonal specifically detects PGM3 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen PGM3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1A11
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGM1PAGMAcetylglucosamine phosphomutase, DKFZP434B187, EC 5.4.2, EC 5.4.2.3, FLJ11614, FLJ13623, N-acetylglucosamine-phosphate mutase, N-acetylglucosamine-phosphate mutase 1, PGM 3, phosphoacetylglucosamine mutase, phosphoglucomutase 3, Phosphoglucomutase-3
Gene Symbols PGM3
Host Species Mouse
Immunogen Full length recombinant protein of human PGM3 (NP_056414) produced in HKE293T cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5238.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.