Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PHA 665752

Catalog No. p-4784281
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective MET inhibitor

Potent, selective and ATP-competitive inhibitor of MET kinase (IC50 values are 9, 68, 200, 1400, 3000, 3800 and 6000 nM for MET, Ron, Flk-1, c-abl, FGFR1, EGFR and c-src respectively and > 10000 nM for IGF-IR, PDGFR, AURORA2, PKA, PKBα, p38α, MK2 and MK3). Antitumor agent; inhibits tumorigenicity and angiogenesis in mouse lung cancer xenografts.

Chemical Identifiers

CAS 477575-56-7
Molecular Formula C32H34Cl2N4O4S

Specifications

Quantity 10 mg
Formula Weight 641.61
Purity >98%
Molecular Weight (g/mol) 641.608
Chemical Name or Material PHA 665752
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.