Learn More
Promega pHAb Thiol Reactive Dye
Catalog No. p-7068010
Click to view available options
Quantity:
250 μg
4 x 250 μg
pHAb Amine Reactive Dye exhibits very low fluorescence at pH > 7 and much higher fluorescence as the pH of the solution becomes acidic. pHAb Dyes are designed specifically for antibody labeling.
  • Measure Antibody Internalization in Real Time
  • Compatible with 96-well, plate-based assay
  • High signal-to-background ratios
  • On-Bead conjugation
Specifications
Quantity 4 x 250 μg
Content And Storage -30° to -10°C for 1 month and <-65°C for long-term storage
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.