Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Phalloidin-Janelia Fluor 646
Click to view available options
:
12 μg
Description
Phalloidin-Janelia Fluor® 646 is a red fluorescent F-actin probe. Phalloidin-Janelia Fluor® 646 is composed of the F-actin probe, Phalloidin (Cat. No. 4535), conjugated to Janelia Fluor® 646 (Cat. No. 6148). It is suitable for use in confocal and super-resolution microscopy techniques such as dSTORM. Excitation/emission maximum λ = 646/664 nm.
Specifications
Specifications
Quantity | 12 μg |
Target | Janelia Fluor Dyes |
Molecular Formula | C64H75N11O13SSi |
Purity | 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction