Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phantom Antibody, Novus Biologicals™
SDP

Catalog No. NB300658005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300658005 0.05 mg
NB018165 0.025 mg
NB300658 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300658005 Supplier Novus Biologicals Supplier No. NB3006580.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phantom Polyclonal specifically detects Phantom in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Phantom
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias Exo, Exocephalon
Host Species Rabbit
Immunogen Synthetic peptide for Saxophone antibodies are (WIT-101AP XXXXXXXXXXXXXXXXXX and; WIT-112APXXXXXXXXXXXXXXXXXXXXXXX; All peptides were amidated for conjugation.
Quantity 0.05 mg
Primary or Secondary Primary
Test Specificity Phantom
Target Species Drosophila
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.