Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PHCCC
GREENER_CHOICE

Catalog No. p-4785332
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent group I mGlu antagonist

Group I metabotropic glutamate receptor antagonist (IC50 ∽ 3 μM); 67 times more potent than (S)-4-carboxyphenylglycine (Cat. No. 0323). Also a positive allosteric modulator for mGlu4a; potentiates L-AP4-mediated inhibition of striatopallidal synaptic transmission in vitro. Displays antiParkinsonian effects in rats in vivo.

Chemical Identifiers

CAS 177610-87-6
Molecular Formula C17H14N2O3

Specifications

Quantity 10 mg
Formula Weight 294.31
Purity >98%
Molecular Weight (g/mol) 294.31
Chemical Name or Material PHCCC
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.