Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD4/HIF Prolyl Hydroxylase 4 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB100295F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100295F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100295F Supplier Novus Biologicals Supplier No. NB100295F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHD4/HIF Prolyl Hydroxylase 4 Polyclonal specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human, Mouse (Negative), Rat samples. It is validated for Electron Microscopy, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen PHD4/HIF Prolyl Hydroxylase 4
Applications Electron Microscopy, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_808808.1
Gene Alias EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase
Gene Symbols P4HTM
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681).
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54681
Test Specificity Human. Does not appear to work in Mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
Target Species Human, Mouse (Negative), Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.