Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenanthrene, tech. 90%
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid
Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid
Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 97°C to 101°C |
| Density | 0.98 |
| Boiling Point | 340°C |
| Flash Point | 171°C (339°F) |
| Quantity | 100 g |
| UN Number | UN3077 |
| Beilstein | 1905428 |
| Merck Index | 14,7212 |
| Solubility Information | Soluble in alcohol, benzene, toluene, and glacial acetic acid |
| Formula Weight | 178.23 |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 201-581-5 |
| RTECSNumber | SF7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction