Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phenol Red sodium salt

Catalog No. p-7026127
Click to view available options
Quantity:
5 g
25 g

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

Phenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.

Solubility
Soluble in water yielding a dark red solution (25 mg/ml), alcohol (4 mg/ml), methanol, ethanol (200 mg/ml), and aqueous alkali hydroxides or carbonates with formation of a red colored solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
Synonym phenol red, sodium salt, phenolsulfonephthalein sodium salt, sodium 4-3-4-hydroxyphenyl-1,1-dioxido-3h-benzo c 1,2 oxathiol-3-yl phenolate, phenol red sodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, sodium salt 1:1, phenol red, acs, phenolredsodiumsalt, phenolsulfonephthalein sodium, sodium 4-3-4-hydroxyphenyl-1,1-dioxo-2,1??-benzoxathiol-3-yl benzenolate
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285°C (decomposition)
Quantity 5 g
Beilstein 6260026
Merck Index 14,7247
Solubility Information Soluble in water yielding a dark red solution (25mg/ml),alcohol (4mg/ml),methanol,ethanol (200mg/ml),and aqueous alkali hydroxides or carbonates with formation of a red colored solution.
Formula Weight 376.37
Chemical Name or Material Phenol Red sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-057-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.