Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenyl isocyanate, 99%
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -31.3°C |
| Color | Colorless to Yellow |
| Density | 1.0950g/mL |
| Boiling Point | 162.0°C to 163.0°C |
| Flash Point | 51°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C7H5NO |
| Refractive Index | 1.5350 to 1.5370 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. Fatal if inhaled. May cause respiratory irritation. May cause an allergic skin reaction. Causes severe skin burns and eye damage.<br |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call |
| EINECSNumber | 203-137-6 |
| RTECSNumber | DA3675000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction