Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl trans-beta-styryl sulfone, 96%

Catalog No. p-7024230
Click to view available options
Quantity:
1 g
5 g

CAS: 16212-06-9 | C14H12O2S | 244.308 g/mol

Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16212-06-9
Molecular Formula C14H12O2S
Molecular Weight (g/mol) 244.308
MDL Number MFCD00159177
InChI Key DNMCCXFLTURVLK-VAWYXSNFSA-N
Synonym phenyl trans-styryl sulfone, phenyl styryl sulfone, sulfone, phenyl styryl, phenyl trans-styryl sulphone, phenyl 2-phenylvinyl sulfone, benzene, 2-phenylethenyl sulfonyl, e-2-benzenesulfonyl ethenyl benzene, 1e-2-phenylvinyl sulfonyl benzene, styrylsulfonylbenzene, e-2-phenylethenyl sulphonylbenzene
PubChem CID 736143
IUPAC Name [(E)-2-(benzenesulfonyl)ethenyl]benzene
SMILES C1=CC=C(C=C1)C=CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Formula Weight 244.32
Percent Purity 98%
Chemical Name or Material Phenyl trans-beta-styryl sulfone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.