Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylalanine Hydroxylase Antibody (17D10), Novus Biologicals™
SDP

Catalog No. NB298091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298091 100 μL
NB298090 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298091 Supplier Novus Biologicals Supplier No. NBP326443100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Phenylalanine Hydroxylase Monoclonal antibody specifically detects Phenylalanine Hydroxylase in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Phenylalanine Hydroxylase
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 17D10
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 1.14.16, EC 1.14.16.1, PH, phe-4-monooxygenase, phenylalanine 4-monooxygenase, phenylalanine hydroxylase, phenylalanine-4-hydroxylase, PKU, PKU1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Phenylalanine Hydroxylase [UniProt P00439]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5053
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.