Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenylalanine Hydroxylase Antibody (17D10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Phenylalanine Hydroxylase |
---|---|
Clone | 17D10 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Phenylalanine Hydroxylase Monoclonal antibody specifically detects Phenylalanine Hydroxylase in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Phenylalanine Hydroxylase | |
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5053 | |
IgG | |
Affinity purified |
17D10 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
EC 1.14.16, EC 1.14.16.1, PH, phe-4-monooxygenase, phenylalanine 4-monooxygenase, phenylalanine hydroxylase, phenylalanine-4-hydroxylase, PKU, PKU1 | |
A synthesized peptide derived from Human Phenylalanine Hydroxylase [UniProt P00439] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title