Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF14 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017291 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017291 Supplier Novus Biologicals Supplier No. NBP281980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHF14 Polyclonal specifically detects PHF14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PHF14
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias KIAA0783, MGC176640, PHD finger protein 14
Gene Symbols PHF14
Host Species Rabbit
Immunogen PHF14 antibody was raised against a 19 amino acid peptide near the amino terminus of human PHF14. The immunogen is located within amino acids 210 - 260 of PHF14 . Amino Acid Squence: KRKGRSASQKEGSDGDNED
Molecular Weight of Antigen 100 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9678
Test Specificity PHF14 antibody is human, mouse and rat reactive. Multiple isoforms of PHF14 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.