Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF16 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB362716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362716 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362716 Supplier Novus Biologicals Supplier No. NBP204021DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHF16 Polyclonal antibody specifically detects PHF16 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PHF16
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias MGC138748, MGC138749, PHD finger protein 16KIAA0215JADE3, protein Jade-3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 773 and 823 of human PHD Finger Protein 16 using the numbering given in entry NP_055550.1 (GeneID 9767).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9767
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.