Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF21B Antibody (OTI3D1), HRP, Novus Biologicals™
SDP

Catalog No. NBP273367H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273367H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273367H Supplier Novus Biologicals Supplier No. NBP273367H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PHF21B Monoclonal specifically detects PHF21B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PHF21B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3D1
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias BHC80L, FLJ34161, KIAA1661, PHD finger protein 21B, PHD finger protein 4, PHF4
Gene Symbols PHF21B
Host Species Mouse
Immunogen Full length recombinant protein of human PHF21B (NP_612424) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 112885.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.