Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF21B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576286
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA576286 100 μL
1 options

Catalog No. PIPA576286

Supplier: Invitrogen™ PA576286

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

PHF21B (PHD finger protein 21B), also known as PHF4, is a 531 amino acid protein that contains one PHD-type zinc finger and is expressed as multiple alternatively spliced isoforms. The gene encoding PHF21B maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF21B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF21B
Gene Accession No. Q8C966, Q96EK2
Gene Alias A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739
Gene Symbols PHF21B
Host Species Rabbit
Immunogen Recombinant fusion protein of human PHF21B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112885, 271305, 300117
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.