Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF8 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093314 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093314 Supplier Novus Biologicals Supplier No. NB10093314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

PHF8 Polyclonal specifically detects PHF8 in Human samples. It is validated for Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen PHF8
Applications Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation, KnockDown Validated
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9UPP1
Gene Alias EC 1.14.11.27, histone lysine demethylase PHF8, jumonji C domain-containing histone demethylase 1F, KIAA1111ZNF422JHDM1F, MRXSSD, PHD finger protein 8DKFZp686E0868
Gene Symbols PHF8
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human PHD finger protein 8 using the numbering given in entry CAI41576.1 (GeneID 23133).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23133
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, and White-tufted-ear marmoset.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.