Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHGDH Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB350537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350537 Supplier Novus Biologicals Supplier No. NBP318850PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHGDH Polyclonal antibody specifically detects PHGDH in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PHGDH
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 3,3-phosphoglycerate dehydrogenase, 3PGDH, 3-PGDH, D-3-phosphoglycerate dehydrogenase, EC 1.1.1, EC 1.1.1.95, MGC3017, PDG, PGAD, PGD, PGDH, phosphoglycerate dehydrogenase, SERA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PHGDH fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26227
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.