Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Philanthotoxin 74, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
AMPA receptor antagonist. Non-selective homomeric GluA3 and GluA1 receptor antagonist (IC50 values are 263 and 296 nM respectively). Inhibits homomeric GluA3 and GluA1 with a 100-fold greater potency than heteromeric GluA1/2 and GluA2/3 receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 507.54 |
| Percent Purity | >98% |
| Chemical Name or Material | Philanthotoxin 74 |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction