Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix, 1,250 reactions PROMO
Click to view available options
:
Thermo Scientific™ PCR Master Mix
Description
Phire Plant Direct PCR Master Mix has been tested with leaves and seeds from a wide variety of plant species as well as fungi, bacteria, and yeast. The kit includes a complete set of optimized reagents and detailed protocols, making it an ideal choice for amplification of plant DNA.Suitable for 1250 reactions.
Features
- Direct PCR allows sample to be added directly to PCR reaction, reducing time-consuming and expensive DNA purification steps
- Available Dilution & Storage protocol allows several PCR reactions from one sample
- Specially engineered Phire Hot Start II DNA Polymerase for extremely short PCR protocol times
- Master mix format with premixed gel loading dye minimizes risk of cross-contamination, reduces sample handling, and allows direct loading on gel
Applications
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
Notes
- Annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | 2X Phire Plant Direct PCR Master Mix, 10 x 1.25 mL, Dilution Buffer, 2 x 5 mL, 25 μM Control Primer Mix, 40 μL, Nuclease-free Water, 10 x 1.25 mL, O'GeneRuler Express DNA Ladder, 50 μg (Sufficient for 100 Applications). Store at -20°C. |
| GC-Rich PCR Performance | Medium |
| PCR Method | Direct PCR |
| Polymerase | Phire Hot Start II DNA Polymerase |
| Reaction Speed | Fast |
| Product Type | Direct PCR Master Mix |
| Quantity | 1250 Reactions |
| Shipping Condition | Wet or Dry Ice |
| For Use With (Application) | Genotyping, Hot-start PCR |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction