Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix PROMO
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix.
Supplier: Thermo Scientific™ F160S
Description
Highlights
- Direct PCR - sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
- Dilution and Storage protocol available - allows tens of PCR reactions from one tiny sample and allows re-testing
- Specially engineered Phire Hot Start II DNA polymerase - extremely short PCR protocol times
- Master Mix format with premixed gel loading dye - minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Applications
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
Includes
- 2x Phire Plant Direct PCR Master Mix
- Dilution Buffer
- Control Primer Mix (25 μM each)
- Water, nuclease- free
- O'GeneRuler Express DNA Ladder
Specifications
Contains: 2 x 1.25 mL Phire Plant Direct PCR Master Mix (2X), 5 mL Dilution Buffer, 40 μL Control Primer Mix (25 μM each), 2 x 1.25 mL Water, nuclease- free, 50 μg O'GeneRuler Express DNA Ladder (sufficient for 100 applications), Store at -20°C. | |
Direct PCR | |
Fast | |
Phire | |
250 x 50 μL Reactions | |
Built-In Hot Start | |
Blunt | |
Plant |
Medium | |
Phire Hot Start II DNA polymerase | |
Yes | |
Direct PCR Master Mix | |
Genotyping | |
250 x 50 L Reactions | |
SuperMix or Master Mix | |
DNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction